Learn More
Biolegend APC/FireTM 750 anti-mouse CD45
Catalog No. 502722643
Encompass_Preferred
APC/Fire™ 750 anti-mouse CD45 [I3/2.3]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-272-2643 Supplier Biolegend Supplier No. 147713
APC/Fire™ 750 anti-mouse CD45 [I3/2.3]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 25 μg