Learn More
Biolegend APC/FireTM 810 anti-human CD9
Catalog No. 502722450
Encompass_Preferred
APC/Fire™ 810 anti-human CD95 (Fas) [DX2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-272-2450 Supplier Biolegend Supplier No. 305663
APC/Fire™ 810 anti-human CD95 (Fas) [DX2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests