Learn More
Cytek Biosciences Inc APC aMu CD49b Anti Primary
Supplier: Cytek Biosciences Inc 200491U100
Product Name: APC Anti-Mouse CD49b (HMa2)
Product Class: Antibody
Type: Primary
Product Name: APC Anti-Mouse CD49b (HMa2)
Cytek Catalog Number: 20-0491-U100
Marker: CD49b
Target Species: Mouse
Clonality: Monoclonal
Clone: HMa2
Host Species: Armenian Hamster
Isotype: IgG
Conjugation: APC
Size: 100 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.