Learn More
ACROBiosystems APC-Labeled Human HLA-A0201
Catalog No. 502534778
Encompass_Preferred
APC-Labeled Human HLA-A*0201B2MNY-ESO-1 (SLLMWITQV) Tetramer Protein (HL1-HA2H8) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) SLLMWITQV peptide (Accession # AAA59606.1 (HLA-A*0201) P61769-1 (B2M) SLLMWITQV).
Catalog No. 50-253-4778 Supplier ACROBiosystems Supplier No. HL1HA2H825TESTS
APC-Labeled Human HLA-A*0201B2MNY-ESO-1 (SLLMWITQV) Tetramer Protein (HL1-HA2H8) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) SLLMWITQV peptide (Accession # AAA59606.1 (HLA-A*0201) P61769-1 (B2M) SLLMWITQV).