Learn More
ACROBiosystems APC-Labeled Monoclonal Anti-G4S Linker Antibody (Rabbit IgG, Clone 016) - 25 tests
Catalog No. NC3432244
Encompass_Preferred
APC-conjugated monoclonal antibody that binds the G4S linker tag and is formulated for flow cytometry detection of G4S-tagged recombinant proteins. Supplied for 25 tests in a formulation containing 0.03% Proclin preservative. Host: rabbit IgG; clone: 016.
  • APC conjugate optimized for flow cytometry.
  • Recognizes the G4S linker tag in recombinant constructs.
  • Host rabbit IgG, clone 016.
  • Supplied for 25 flow cytometry tests.
  • Formulated with 0.03% Proclin preservative.
  • Validated for flow cytometry applications.
Catalog No. NC3432244 Supplier ACROBiosystems Supplier No. G4SAFMY2525TESTS
APC-conjugated monoclonal antibody that binds the G4S linker tag and is formulated for flow cytometry detection of G4S-tagged recombinant proteins. Supplied for 25 tests in a formulation containing 0.03% Proclin preservative. Host: rabbit IgG; clone: 016.
  • APC conjugate optimized for flow cytometry.
  • Recognizes the G4S linker tag in recombinant constructs.
  • Host rabbit IgG, clone 016.
  • Supplied for 25 flow cytometry tests.
  • Formulated with 0.03% Proclin preservative.
  • Validated for flow cytometry applications.