Learn More
Cytek Biosciences Inc APC MS CD11C (N418) 100UG
Supplier:  Cytek Biosciences Inc 200114U100
Encompass_Preferred
APC Anti-Mouse CD11c (N418) Isotype: Armenian Hamster IgG Apps: FC Size: 100 ug
Catalog No. 50-105-3054