Learn More
Cytek Biosciences Inc APC MS CD4 (GK1.5) 100UG
Supplier:  Cytek Biosciences Inc 200041U100
Encompass_Preferred
APC Anti-Mouse CD4 (GK1.5) Isotype: Rat IgG2b, kappa Apps: FC Size: 100 ug
Catalog No. 50-105-3018