Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc APC aMu CD45.1 Anti Primary

Catalog No. 502014145
Encompass_Preferred

Product Name: APC Anti-Mouse CD45.1 (A20)
Product Class: Antibody
Type: Primary
Product Name: APC Anti-Mouse CD45.1 (A20)
Cytek Catalog Number: 20-0453-U100
Marker: CD45.1
Target Species: Mouse
Clonality: Monoclonal
Clone: A20
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: APC
Size: 100 μg

Catalog No. 50-201-4145 Supplier Cytek Biosciences Inc Supplier No. 200453U100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Product Name: APC Anti-Mouse CD45.1 (A20)
Product Class: Antibody
Type: Primary
Product Name: APC Anti-Mouse CD45.1 (A20)
Cytek Catalog Number: 20-0453-U100
Marker: CD45.1
Target Species: Mouse
Clonality: Monoclonal
Clone: A20
Host Species: Mouse
Isotype: IgG2a, k
Conjugation: APC
Size: 100 μg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.