Learn More
Cytek Biosciences Inc APC MS CD45.2 (104) 100UG
Supplier:  Cytek Biosciences Inc 200454U100
Encompass_Preferred
APC Anti-Mouse CD45.2 (104) Isotype: Mouse IgG2a, kappa Apps: FC Size: 100 ug
Catalog No. 50-105-3096