Learn More
Cytek Biosciences Inc APC MS CD45 (30-F11) 25UG
Supplier:  Cytek Biosciences Inc 200451U025
Encompass_Preferred
APC Anti-Mouse CD45 (30-F11) Isotype: Rat IgG2b, kappa Apps: FC Size: 25 ug
Catalog No. 50-105-3089