Learn More
Cytek Biosciences Inc APC MS IFN G (XMG1.2) 100UG
Supplier:  Cytek Biosciences Inc 207311U100
Encompass_Preferred
APC Anti-Mouse IFN gamma (XMG1.2) Isotype: Rat IgG1, kappa Apps: FC Size: 100 ug
Catalog No. 50-105-3171