Learn More
Biolegend APC anti-mouse H-2Kb bound to SIINFEKL
Supplier:  Biolegend 141606
Encompass_Preferred
APC anti-mouse H-2Kb bound to SIINFEKL [25-D1.16]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC, ICC; Size: 100 ug
Catalog No. NC1499090