Learn More
Biolegend APC Mouse IgG1, κ isotype Ctrl (FC)
Supplier:  Biolegend 400122
Encompass_Preferred
APC Mouse IgG1, κ isotype Ctrl (FC) [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse ; Apps: FC; Size: 100 tests
Catalog No. 50-168-838