Learn More
Biolegend APC Mouse IgG1, κ Isotype Ctrl (ICFC)
Supplier:  Biolegend 400141
Encompass_Preferred
APC Mouse IgG1, κ Isotype Ctrl (ICFC) [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse ; Apps: ICFC; Size: 25 tests
Catalog No. 50-168-848