Learn More
Biolegend APC Mouse IgG2a, κ isotype Ctrl (FC)
Supplier:  Biolegend 400222
Encompass_Preferred
APC Mouse IgG2a, κ isotype Ctrl (FC) [MOPC-173]; Isotype: Mouse IgG2a, κ; Apps: FC; Size: 100 tests
Catalog No. NC9080952