Learn More
Biolegend APC Mouse IgG2a, κ isotype Ctrl (ICFC)
Supplier:  Biolegend 400246
Encompass_Preferred
APC Mouse IgG2a, κ isotype Ctrl (ICFC) [MOPC-173]; Isotype: Mouse IgG2a, κ; Apps: ICFC; Size: 100 tests
Catalog No. 50-168-881