Learn More
ABclonal Technology APC Rabbit anti-Human CD44 mAb
SDP
Catalog No. 5000005602
Encompass_Preferred
The protein encoded by this gene is a cell-surface glycoprotein involved in cell-cell interactions cell adhesion and migration It is a receptor for hyaluronic acid (HA) and can also interact with other ligands such as osteopontin collagens and matrix metalloproteinases (MMPs) This protein participates in a wide variety of cellular functions including lymphocyte activation recirculation and homing hematopoiesis and tumor metastasis Transcripts for this gene undergo complex alternative splicing that results in many functionally distinct isoforms however the full length nature of some of these variants has not been determined Alternative splicing is the basis for the structural and functional diversity of this protein and may be related to tumor metastasis
Catalog No. 50-000-05602 Supplier ABclonal Technology Supplier No. A24106100T
The protein encoded by this gene is a cell-surface glycoprotein involved in cell-cell interactions cell adhesion and migration It is a receptor for hyaluronic acid (HA) and can also interact with other ligands such as osteopontin collagens and matrix metalloproteinases (MMPs) This protein participates in a wide variety of cellular functions including lymphocyte activation recirculation and homing hematopoiesis and tumor metastasis Transcripts for this gene undergo complex alternative splicing that results in many functionally distinct isoforms however the full length nature of some of these variants has not been determined Alternative splicing is the basis for the structural and functional diversity of this protein and may be related to tumor metastasis