Learn More
ABclonal Technology APC Rabbit anti-Human CD5 mAb
SDP
Catalog No. 5000008878
Encompass_Preferred
This gene encodes a member of the scavenger receptor cysteine-rich (SRCR) superfamily Members of this family are secreted or membrane-anchored proteins mainly found in cells associated with the immune system This protein is a type-I transmembrane glycoprotein found on the surface of thymocytes T lymphocytes and a subset of B lymphocytes The encoded protein contains three SRCR domains and may act as a receptor to regulate T-cell proliferation Alternative splicing results in multiple transcript variants encoding different isoforms
Catalog No. 50-000-08878 Supplier ABclonal Technology Supplier No. A26829100T
This gene encodes a member of the scavenger receptor cysteine-rich (SRCR) superfamily Members of this family are secreted or membrane-anchored proteins mainly found in cells associated with the immune system This protein is a type-I transmembrane glycoprotein found on the surface of thymocytes T lymphocytes and a subset of B lymphocytes The encoded protein contains three SRCR domains and may act as a receptor to regulate T-cell proliferation Alternative splicing results in multiple transcript variants encoding different isoforms