Learn More
ABclonal Technology APC Rabbit anti-/Human CD55
SDP
Supplier:  ABclonal Technology A26739500T
Encompass_Preferred
This gene encodes an inhibitor of both the classical and the alternative pathways of complement activation The encoded preproprotein undergoes post-translational processing to generate a mature polypeptide anchored to the plasma membrane via a glycosylphosphatidylinositol moiety Erythrocytes from mice deficient in the encoded protein exhibit impaired regulation of complement activation resulting in enhanced complement deposition Mice lacking the encoded protein exhibit enhanced susceptibility to experimentally induced myasthenia gravis This gene is located adjacent to a closely related gene on chromosome 1
Catalog No. 50-000-08941