Learn More
ABclonal Technology APC Rabbit anti-Human IL-17A
Supplier: ABclonal Technology A27825200T
This gene encodes a proinflammatory cytokine from the IL-17 receptor family primarily produced by activated T cells IL-17A activates pathways that induce inflammatory molecules chemokines antimicrobial peptides and tissue remodeling proteins It plays key roles in host defense immune modulation and tissue repair by promoting chemokine production and recruiting myeloid cells to sites of inflammation IL-17A also regulates NF- B and MAPK pathways and stimulates expression of IL6 COX-2 and nitric oxide It is implicated in infections autoimmune diseases cancer and contributes to lung damage in severe COVID-19