Learn More
ABclonal Technology APC Rabbit anti-Human IL-17A
SDP
Catalog No. 5000013843
Encompass_Preferred
This gene encodes a proinflammatory cytokine from the IL-17 receptor family primarily produced by activated T cells IL-17A activates pathways that induce inflammatory molecules chemokines antimicrobial peptides and tissue remodeling proteins It plays key roles in host defense immune modulation and tissue repair by promoting chemokine production and recruiting myeloid cells to sites of inflammation IL-17A also regulates NF- B and MAPK pathways and stimulates expression of IL6 COX-2 and nitric oxide It is implicated in infections autoimmune diseases cancer and contributes to lung damage in severe COVID-19
Catalog No. 50-000-13843 Supplier ABclonal Technology Supplier No. A27825500T
This gene encodes a proinflammatory cytokine from the IL-17 receptor family primarily produced by activated T cells IL-17A activates pathways that induce inflammatory molecules chemokines antimicrobial peptides and tissue remodeling proteins It plays key roles in host defense immune modulation and tissue repair by promoting chemokine production and recruiting myeloid cells to sites of inflammation IL-17A also regulates NF- B and MAPK pathways and stimulates expression of IL6 COX-2 and nitric oxide It is implicated in infections autoimmune diseases cancer and contributes to lung damage in severe COVID-19