Learn More
ABclonal Technology APC Rabbit anti- IL1R1/CD121a
SDP
Catalog No. 5000010661
Encompass_Preferred
Enables interleukin-1 binding activity interleukin-1 receptor activity and protease binding activity Involved in interleukin-1-mediated signaling pathway Acts upstream of or within several processes including cytokine-mediated signaling pathway positive regulation of interleukin-1-mediated signaling pathway and positive regulation of neutrophil extravasation Located in external side of plasma membrane Is expressed in brain dorsal aorta liver reproductive system and spleen Used to study type 1 diabetes mellitus Human ortholog(s) of this gene implicated in systemic scleroderma and type 1 diabetes mellitus Orthologous to human IL1R1 (interleukin 1 receptor type 1)
Catalog No. 50-000-10661 Supplier ABclonal Technology Supplier No. A25859500T
Enables interleukin-1 binding activity interleukin-1 receptor activity and protease binding activity Involved in interleukin-1-mediated signaling pathway Acts upstream of or within several processes including cytokine-mediated signaling pathway positive regulation of interleukin-1-mediated signaling pathway and positive regulation of neutrophil extravasation Located in external side of plasma membrane Is expressed in brain dorsal aorta liver reproductive system and spleen Used to study type 1 diabetes mellitus Human ortholog(s) of this gene implicated in systemic scleroderma and type 1 diabetes mellitus Orthologous to human IL1R1 (interleukin 1 receptor type 1)