Learn More
ABclonal Technology APC Rabbit anti-Mouse CD103
SDP
Catalog No. 5000011832
Encompass_Preferred
Predicted to enable metal ion binding activity Predicted to act upstream of or within cell adhesion and integrin-mediated signaling pathway Located in external side of plasma membrane Is expressed in several structures including alimentary system genitourinary system lymph node nasal septum and nucleus pulposus Orthologous to human ITGAE (integrin subunit alpha E)
Catalog No. 50-000-11832 Supplier ABclonal Technology Supplier No. A27134100T
Predicted to enable metal ion binding activity Predicted to act upstream of or within cell adhesion and integrin-mediated signaling pathway Located in external side of plasma membrane Is expressed in several structures including alimentary system genitourinary system lymph node nasal septum and nucleus pulposus Orthologous to human ITGAE (integrin subunit alpha E)