Learn More
ABclonal Technology APC Rabbit anti-Mouse CD11b
SDP
Catalog No. 5000010051
Encompass_Preferred
Enables complement component C3b binding activity heparan sulfate proteoglycan binding activity and heparin binding activity Contributes to cargo receptor activity Involved in several processes including central nervous system development endocytosis and positive regulation of neuron death Acts upstream of or within several processes including activated T cell proliferation leukocyte migration and microglia development Located in external side of plasma membrane and nucleus Is integral component of membrane Part of integrin alphaM-beta2 complex Is expressed in several structures including central nervous system heart lymphatic vessel thymus and yolk sac Human ortholog(s) of this gene implicated in lupus nephritis Orthologous to human ITGAM (integrin subunit alpha M)
Catalog No. 50-000-10051 Supplier ABclonal Technology Supplier No. A26266100T
Enables complement component C3b binding activity heparan sulfate proteoglycan binding activity and heparin binding activity Contributes to cargo receptor activity Involved in several processes including central nervous system development endocytosis and positive regulation of neuron death Acts upstream of or within several processes including activated T cell proliferation leukocyte migration and microglia development Located in external side of plasma membrane and nucleus Is integral component of membrane Part of integrin alphaM-beta2 complex Is expressed in several structures including central nervous system heart lymphatic vessel thymus and yolk sac Human ortholog(s) of this gene implicated in lupus nephritis Orthologous to human ITGAM (integrin subunit alpha M)