Learn More
ABclonal Technology APC Rabbit anti-Mouse CD24 mAb
SDP
Catalog No. 5000010611
Encompass_Preferred
CD24 also know as heat stable antigen HSA is a P-selectin ligand involved in adhesion It is a GPI-anchored glycoprotein expressed on many types of cells including hematopoietic cells neural cells and epithelial cells CD24 is widely used to delineate stages of lymphocyte development It also binds to Siglec-10 in humans or Siglec-G in mice CD24 is frequently used as a marker to identify and isolate cancer stem cells in various cancer types
Catalog No. 50-000-10611 Supplier ABclonal Technology Supplier No. A25890100T
CD24 also know as heat stable antigen HSA is a P-selectin ligand involved in adhesion It is a GPI-anchored glycoprotein expressed on many types of cells including hematopoietic cells neural cells and epithelial cells CD24 is widely used to delineate stages of lymphocyte development It also binds to Siglec-10 in humans or Siglec-G in mice CD24 is frequently used as a marker to identify and isolate cancer stem cells in various cancer types