Learn More
ABclonal Technology APC Rabbit anti-Mouse CD38 mAb
Supplier: ABclonal Technology A2736525UG
This gene encodes a non-lineage-restricted type II transmembrane glycoprotein that synthesizes and hydrolyzes cyclic adenosine 5 -diphosphate-ribose an intracellular calcium ion mobilizing messenger The release of soluble protein and the ability of membrane-bound protein to become internalized indicate both extracellular and intracellular functions for the protein This protein has an N-terminal cytoplasmic tail a single membrane-spanning domain and a C-terminal extracellular region with four N-glycosylation sites Knockout mice deficient for this gene display altered humoral immune responses In addition knockout mice exhibit higher locomotor activity and defects in nurturing and social behaviors