Learn More
ABclonal Technology APC Rabbit anti-Mouse CD38 mAb
SDP
Catalog No. 5000010448
Encompass_Preferred
This gene encodes a non-lineage-restricted type II transmembrane glycoprotein that synthesizes and hydrolyzes cyclic adenosine 5 -diphosphate-ribose an intracellular calcium ion mobilizing messenger The release of soluble protein and the ability of membrane-bound protein to become internalized indicate both extracellular and intracellular functions for the protein This protein has an N-terminal cytoplasmic tail a single membrane-spanning domain and a C-terminal extracellular region with four N-glycosylation sites Knockout mice deficient for this gene display altered humoral immune responses In addition knockout mice exhibit higher locomotor activity and defects in nurturing and social behaviors
Catalog No. 50-000-10448 Supplier ABclonal Technology Supplier No. A27365100UG
This gene encodes a non-lineage-restricted type II transmembrane glycoprotein that synthesizes and hydrolyzes cyclic adenosine 5 -diphosphate-ribose an intracellular calcium ion mobilizing messenger The release of soluble protein and the ability of membrane-bound protein to become internalized indicate both extracellular and intracellular functions for the protein This protein has an N-terminal cytoplasmic tail a single membrane-spanning domain and a C-terminal extracellular region with four N-glycosylation sites Knockout mice deficient for this gene display altered humoral immune responses In addition knockout mice exhibit higher locomotor activity and defects in nurturing and social behaviors