Learn More
ABclonal Technology APC Rabbit anti-Mouse CD47 mAb
SDP
Catalog No. 5000008552
Encompass_Preferred
Predicted to enable protein binding activity involved in heterotypic cell-cell adhesion and thrombospondin receptor activity Involved in regulation of nitric oxide biosynthetic process Acts upstream of or within several processes including monocyte aggregation opsonization and positive regulation of phagocytosis Located in extracellular exosome Is integral component of plasma membrane Is expressed in several structures including alimentary system central nervous system genitourinary system hemolymphoid system gland and liver and biliary system Orthologous to human CD47 (CD47 molecule)
Catalog No. 50-000-08552 Supplier ABclonal Technology Supplier No. A26819500T
Predicted to enable protein binding activity involved in heterotypic cell-cell adhesion and thrombospondin receptor activity Involved in regulation of nitric oxide biosynthetic process Acts upstream of or within several processes including monocyte aggregation opsonization and positive regulation of phagocytosis Located in extracellular exosome Is integral component of plasma membrane Is expressed in several structures including alimentary system central nervous system genitourinary system hemolymphoid system gland and liver and biliary system Orthologous to human CD47 (CD47 molecule)