Learn More
ABclonal Technology APC Rabbit anti-Mouse CD93 mAb
SDP
Catalog No. 5000010371
Encompass_Preferred
Predicted to enable complement component C1q complex binding activity Predicted to be involved in cell-cell adhesion Predicted to act upstream of or within cell adhesion Located in cell surface cytoplasmic vesicle and plasma membrane Is expressed in blood vessel Orthologous to human CD93 (CD93 molecule)
Catalog No. 50-000-10371 Supplier ABclonal Technology Supplier No. A26058200T
Predicted to enable complement component C1q complex binding activity Predicted to be involved in cell-cell adhesion Predicted to act upstream of or within cell adhesion Located in cell surface cytoplasmic vesicle and plasma membrane Is expressed in blood vessel Orthologous to human CD93 (CD93 molecule)