Learn More
ABclonal Technology APC Rat anti-Mouse TER-119 mAb
SDP
Catalog No. 5000010037
Encompass_Preferred
TER-119 is a mouse erythroid lineage-specific monoclonal antibody that reacts with erythroid cells at differentiation stages from early proerythroblast to mature erythrocyte but not with cells showing typical erythroid blast-forming unit (BFU-E) and erythroid colony-forming unit (CFU-E) activities TER-119 recognizes a 52-kDa molecule on erythrocyte membranes TER-119 antigen is a molecule associated with cell-surface glycophorin A but not with glycophorin A itself
Catalog No. 50-000-10037 Supplier ABclonal Technology Supplier No. A27476100T
TER-119 is a mouse erythroid lineage-specific monoclonal antibody that reacts with erythroid cells at differentiation stages from early proerythroblast to mature erythrocyte but not with cells showing typical erythroid blast-forming unit (BFU-E) and erythroid colony-forming unit (CFU-E) activities TER-119 recognizes a 52-kDa molecule on erythrocyte membranes TER-119 antigen is a molecule associated with cell-surface glycophorin A but not with glycophorin A itself