Learn More
Enterprise Technology Solutions APC REPLACEMENT BATTERY CARTRIDGE #154
SDP
Catalog No. 502097107
Encompass_Preferred
The Replacement Battery Cartridge #154 from APC is a replacement battery pack for APC Uninterruptible Power Supplies (UPS). With a lifespan of 2 to 5 years, this maintenance-free, sealed, and leakproof lead-acid battery with suspended electrolytes is hot-swappable and RoHS-compliant. Connected devices are protected by a limited lifetime equipment protection policy and UPS protection is provided by a limited 1-year repair or replace warranty.This battery is compatible with APC's BE600M1, BE670M1, and BN675M1.
Catalog No. 50-209-7107 Supplier Enterprise Technology Solutions Supplier No. APCRBC154
The Replacement Battery Cartridge #154 from APC is a replacement battery pack for APC Uninterruptible Power Supplies (UPS). With a lifespan of 2 to 5 years, this maintenance-free, sealed, and leakproof lead-acid battery with suspended electrolytes is hot-swappable and RoHS-compliant. Connected devices are protected by a limited lifetime equipment protection policy and UPS protection is provided by a limited 1-year repair or replace warranty.This battery is compatible with APC's BE600M1, BE670M1, and BN675M1.