Learn More
Enterprise Technology Solutions APC REPLACEMENT BATTERY CARTRIDGE #4 - UPS BATTERY - LEAD ACID
SDP
Catalog No. 502097162
Encompass_Preferred
APC RBC4. Battery technology: Sealed Lead Acid (VRLA). Dimensions (WxDxH): 3.9 x 3.7 x 5.9" (99.1 x 94 x 149.9 mm). Weight: 8.11 lbs (3.68 kg)
Catalog No. 50-209-7162 Supplier Enterprise Technology Solutions Supplier No. RBC4
APC RBC4. Battery technology: Sealed Lead Acid (VRLA). Dimensions (WxDxH): 3.9 x 3.7 x 5.9" (99.1 x 94 x 149.9 mm). Weight: 8.11 lbs (3.68 kg)