Learn More
Enterprise Technology Solutions APC REPLACEMENT BATTERY CARTRIDGE #7 - UPS BATTERY - 1 X LEAD ACID
SDP
Catalog No. 502097166
Encompass_Preferred
APC RBC7. Battery technology: Sealed Lead Acid (VRLA), Product color: Black. Dimensions (WxDxH): 6 x 7.2 x 6.8" (152.4 x 182.9 x 172.7 mm). Weight: 25.8 lbs (11.7 kg)
Catalog No. 50-209-7166 Supplier Enterprise Technology Solutions Supplier No. RBC7
APC RBC7. Battery technology: Sealed Lead Acid (VRLA), Product color: Black. Dimensions (WxDxH): 6 x 7.2 x 6.8" (152.4 x 182.9 x 172.7 mm). Weight: 25.8 lbs (11.7 kg)