Learn More
Custom Networks Inc APC Replacement Battery Cartridge, 12V 5Ah, Lead-Acid
SDP
Catalog No. NC3978167
Encompass_Preferred
This Replacement Battery Cartridge is specifically designed to be fully compatible with Smart-UPS on-line systems. It features a 12V 5Ah VRLA battery that is thoroughly tested to restore UPS performance to original specifications. The cartridge includes a 2-year return or replaceable warranty.
  • Brand new battery shipped from strictly managed warehouse.
  • Leak-proof sealed lead-acid battery.
  • Fully assembled and ready for use.
  • Supports authorized recycling for earth-friendly disposal.
  • Designed to be fully compatible with Smart-UPS on-line systems.
Catalog No. NC3978167 Supplier Custom Networks Inc Supplier No. APCRBC152
This Replacement Battery Cartridge is specifically designed to be fully compatible with Smart-UPS on-line systems. It features a 12V 5Ah VRLA battery that is thoroughly tested to restore UPS performance to original specifications. The cartridge includes a 2-year return or replaceable warranty.
  • Brand new battery shipped from strictly managed warehouse.
  • Leak-proof sealed lead-acid battery.
  • Fully assembled and ready for use.
  • Supports authorized recycling for earth-friendly disposal.
  • Designed to be fully compatible with Smart-UPS on-line systems.