Learn More
Custom Networks Inc APC Smart-UPS C, Line Interactive, 1440 VA, Rackmount 2U, 120 V
SDP
Catalog No. NC3978176
Encompass_Preferred
This line-interactive uninterruptible power supply (UPS) is designed to maintain business uptime and continuity, providing conditioned AC power to connected equipment. It features automatic voltage regulation (AVR) and a SmartConnect port for remote monitoring.
  • 1440 VA / 900 W capacity.
  • Rackmount 2U form factor.
  • 120 V input (NEMA 5-15P) and six NEMA 5-15R outlets.
  • SmartConnect port, USB, and serial communication.
  • Graphic LCD interface.
  • Automatic Voltage Regulation (AVR).
  • Cloud-based remote monitoring and firmware upgrades.
  • Efficient green mode operation.
  • Integrated surge protection.
  • User-replaceable lead-acid battery.
  • Predictive battery management.
  • 2-year warranty.
Catalog No. NC3978176 Supplier Custom Networks Inc Supplier No. SMC15002UC
This line-interactive uninterruptible power supply (UPS) is designed to maintain business uptime and continuity, providing conditioned AC power to connected equipment. It features automatic voltage regulation (AVR) and a SmartConnect port for remote monitoring.
  • 1440 VA / 900 W capacity.
  • Rackmount 2U form factor.
  • 120 V input (NEMA 5-15P) and six NEMA 5-15R outlets.
  • SmartConnect port, USB, and serial communication.
  • Graphic LCD interface.
  • Automatic Voltage Regulation (AVR).
  • Cloud-based remote monitoring and firmware upgrades.
  • Efficient green mode operation.
  • Integrated surge protection.
  • User-replaceable lead-acid battery.
  • Predictive battery management.
  • 2-year warranty.