Learn More
Enterprise Technology Solutions APC Smart-UPS SRT 3kVA Rackmount Battery Backup & Surge Protector (208/230V IEC)
SDP
Supplier:  Enterprise Technology Solutions SRT3000RMXLWIEC
Encompass_Preferred
Protect your high-end network equipment and servers from unexpected power outages with the Smart-UPS SRT 3kVA Rackmount Battery Backup & Surge Protector from APC. This 2U rackmount UPS is designed to deliver 2700W / 3000 VA power output at 230V to six IEC 320 C13 and two IEC 320 C19 outlets. It can run for up to about four minutes when at full capacity, providing pure sine-wave power with a double-conversion topography for zero transfer time.
Catalog No. 50-001-73517
Add to Cart
Request a Quote