Learn More
Enterprise Technology Solutions APC Smart-UPS SRT 48V 1kVA and 1.5kVA RM Battery Pack
SDP
Catalog No. 5000173515
Encompass_Preferred
The Smart-UPS SRT 48V 1kVA and 1.5kVA RM Battery Pack from APC serves as an external battery pack for the APC SRT1000RMXLA, SRT1000RMXLA-NC, SRT1500RMXLA, and SRT1500RMXLA-NC Smart-UPS SRT units. The two integrated lead-acid batteries are maintenance-free, leakproof, sealed, and hot-swappable. This rack-mounted, 2U-sized pack features an output voltage of 48V and has an expected battery life of 3-5 years. Protection is provided by a limited 2-year repair or replace warranty.
Catalog No. 50-001-73515 Supplier Enterprise Technology Solutions Supplier No. SRT48RMBP
The Smart-UPS SRT 48V 1kVA and 1.5kVA RM Battery Pack from APC serves as an external battery pack for the APC SRT1000RMXLA, SRT1000RMXLA-NC, SRT1500RMXLA, and SRT1500RMXLA-NC Smart-UPS SRT units. The two integrated lead-acid batteries are maintenance-free, leakproof, sealed, and hot-swappable. This rack-mounted, 2U-sized pack features an output voltage of 48V and has an expected battery life of 3-5 years. Protection is provided by a limited 2-year repair or replace warranty.