Learn More
Custom Networks Inc APC Smart-UPS X, Line Interactive UPS, 2000VA, Rack/Tower 4U, 100-127V, Short Depth
SDP
Catalog No. NC3978223
Encompass_Preferred
The Smart-UPS X is a high-performance 2000VA/1800W line interactive UPS, ideal for IT managers and network administrators. This versatile 4U UPS supports critical electronics such as servers and routers. Its short depth design optimizes space, and it features a user-selectable voltage range of 100V-127V with pure sine wave output.
  • 1920VA/1800W line interactive pure sine wave UPS.
  • Rack/tower convertible 4U design.
  • 100V-127V user-selectable voltage.
  • Expandable runtime with external battery packs.
  • Graceful shutdown via serial or USB.
  • Multiple NEMA output: 6x 5-15R, 3x 5-20R, 1x L5-20R.
  • SmartSlot for optional Network Management Card.
  • 3-year UPS, 2-year battery warranty.
  • User-replaceable battery.
  • Emergency Power Off (EPO) feature.
Catalog No. NC3978223 Supplier Custom Networks Inc Supplier No. SMX2000LV
The Smart-UPS X is a high-performance 2000VA/1800W line interactive UPS, ideal for IT managers and network administrators. This versatile 4U UPS supports critical electronics such as servers and routers. Its short depth design optimizes space, and it features a user-selectable voltage range of 100V-127V with pure sine wave output.
  • 1920VA/1800W line interactive pure sine wave UPS.
  • Rack/tower convertible 4U design.
  • 100V-127V user-selectable voltage.
  • Expandable runtime with external battery packs.
  • Graceful shutdown via serial or USB.
  • Multiple NEMA output: 6x 5-15R, 3x 5-20R, 1x L5-20R.
  • SmartSlot for optional Network Management Card.
  • 3-year UPS, 2-year battery warranty.
  • User-replaceable battery.
  • Emergency Power Off (EPO) feature.