Learn More
Enterprise Technology Solutions APC Symmetra LX Battery Module
SDP
Catalog No. 5000173617
Encompass_Preferred
With a battery life expectancy of 3 to 5 years, the Symmetra LX Battery Module from APC is designed to offer consistent performance making it a reliable power source. The maintanence-free sealed lead-acid battery features an enclosed battery cartridge with 10 battery blocks per string and a capacity of 1,080 volt-amp-hours. Hot pluggable operation makes for easy installation in a variety of configurations including 1U rack mount enclosures.
Catalog No. 50-001-73617 Supplier Enterprise Technology Solutions Supplier No. SYBT5
With a battery life expectancy of 3 to 5 years, the Symmetra LX Battery Module from APC is designed to offer consistent performance making it a reliable power source. The maintanence-free sealed lead-acid battery features an enclosed battery cartridge with 10 battery blocks per string and a capacity of 1,080 volt-amp-hours. Hot pluggable operation makes for easy installation in a variety of configurations including 1U rack mount enclosures.