Learn More
Custom Networks Inc APC Symmetra LX Extended Run Rack-mount, 9 SYBT5 Battery Modules, 208V
SDP
Catalog No. NC3978301
Encompass_Preferred
This Symmetra LX Extended Runtime Battery is equipped with nine battery modules and operates at 208V. Designed for high availability and scalability, it’s ideal for critical power applications. It features a 13U rack-mountable form factor for efficient space use and includes lead-acid batteries from Panasonic. The unit supports an operating temperature range from 0°C to 40°C and functions at altitudes up to 10,000 ft.
  • Connects to a UPS or existing XR frame for extended runtime during power outages.
  • Pre-populated with 9 battery modules.
  • Rackmount form factor with 13U rack height.
  • Daisy chain multiple XR frames for additional runtime.
  • Lead-acid battery type.
  • Designed for high availability and scalability.
  • Supports an operating temperature range from 0°C to 40°C.
  • Functions at altitudes up to 10,000 ft.
Catalog No. NC3978301 Supplier Custom Networks Inc Supplier No. SYARMXR9B9
This Symmetra LX Extended Runtime Battery is equipped with nine battery modules and operates at 208V. Designed for high availability and scalability, it’s ideal for critical power applications. It features a 13U rack-mountable form factor for efficient space use and includes lead-acid batteries from Panasonic. The unit supports an operating temperature range from 0°C to 40°C and functions at altitudes up to 10,000 ft.
  • Connects to a UPS or existing XR frame for extended runtime during power outages.
  • Pre-populated with 9 battery modules.
  • Rackmount form factor with 13U rack height.
  • Daisy chain multiple XR frames for additional runtime.
  • Lead-acid battery type.
  • Designed for high availability and scalability.
  • Supports an operating temperature range from 0°C to 40°C.
  • Functions at altitudes up to 10,000 ft.