Learn More
Custom Networks Inc APC Symmetra LX Extended Run Tower, 9 Battery Modules, Lead-Acid, Black
SDP
Catalog No. NC3978303
Encompass_Preferred
This extended runtime battery is designed for high-speed applications, providing additional runtime in case of a power outage. It is equipped with nine battery modules and includes an installation guide and user manual.
  • Connects to an existing extended run frame to gain more runtime in case of a power outage.
  • Comes pre-populated with 9 battery modules.
  • Tower form factor.
  • Can daisy chain multiple extended run frames for even more runtime.
  • Lead-acid battery type.
  • Includes an installation guide and user manual.
Catalog No. NC3978303 Supplier Custom Networks Inc Supplier No. SYAXR9B9
This extended runtime battery is designed for high-speed applications, providing additional runtime in case of a power outage. It is equipped with nine battery modules and includes an installation guide and user manual.
  • Connects to an existing extended run frame to gain more runtime in case of a power outage.
  • Comes pre-populated with 9 battery modules.
  • Tower form factor.
  • Can daisy chain multiple extended run frames for even more runtime.
  • Lead-acid battery type.
  • Includes an installation guide and user manual.