Learn More
Cole-Parmer APC1667 Amber VOA Vials, 40 mL, Open-Top Cap, 5 mg Ammonium Chloride; 72/cs
Catalog No. NC1823681 Shop All Cole-Parmer Products
Encompass
After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.
Catalog No. NC1823681 Supplier Cole-Parmer Supplier No. 3520718
After collection, samples may require a chemical preservative to extend the holding time to a workable timeframe. Save time by choosing one that already comes premeasured and filled! Preserved vials include the chemical and lot number on the label, and are packed in protective foam blocks.