Learn More
Aviva Systems Biology Corp APC2 Antibody - middle region (ARP50526_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50526_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human APC2. Alternate names: APCLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-60365