Learn More
Cytek Biosciences Inc APC-Cy7 aHu Mu CD45R Anti
Supplier: Cytek Biosciences Inc 250452U100
Product Name: APC-Cyanine7 Anti-Human/Mouse CD45R (B220) (RA3-6B2)
Product Class: Antibody
Type: Primary
Product Name: APC-Cyanine7 Anti-Human/Mouse CD45R (B220) (RA3-6B2)
Cytek Catalog Number: 25-0452-U100
Marker: CD45R (B220)
Target Species: Human|Mouse
Reported Cross Reactivity: Chimpanzee
Clonality: Monoclonal
Clone: RA3-6B2
Host Species: Rat
Isotype: IgG2a, k
Conjugation: APC-Cyanine7
Size: 100 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.