Learn More
Biolegend APC/Fire™ 750 anti-human CD16
Supplier:  Biolegend 980114
Encompass_Preferred
APC/Fire™ 750 anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 uL
Catalog No. 50-213-2092