Learn More
Perkin Elmer US LLC APCI Corona Lens with Needle
Catalog No. 502113065
Encompass_Preferred
For use with Atmospheric Pressure Chemical Ionization (APCI) ion source in with Flexar SQ300 and AxION 2 TOF MS.
Catalog No. 50-211-3065 Supplier Perkin Elmer US LLC Supplier No. MZ301193
For use with Atmospheric Pressure Chemical Ionization (APCI) ion source in with Flexar SQ300 and AxION 2 TOF MS.