Learn More
Waters Corp Waters APCI IonSABRE Probe Heater Assembly, Higher Power, Max 650°C
Catalog No. 50793343 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The APCI IonSABRE Probe Heater Assembly is a component of the IonSABRE APCI system, an HPLC-MS interface for analyzing samples not suited for ElectroSpray ionization. Its higher power design rapidly heats the nebulizer plume, ensuring efficient droplet evaporation for optimal analytical performance.
  • Higher power heater design for atmospheric pressure chemical ionization.
  • Heats nebulizer plume efficiently for rapid droplet evaporation.
  • Supports 100% organic or aqueous mobile phases, 0.2 to 2.0 mL/min.
  • Ideal for standard analytical column LC–MS, normal and reverse phase.
  • APCI probe temperature adjustable up to 650°C.
  • Optimizes temperature for maximum sensitivity.
  • Aids chemical contamination removal from probe tip.
Catalog No. 50-793-343 Supplier Waters Corp Supplier No. M956076BC1
The APCI IonSABRE Probe Heater Assembly is a component of the IonSABRE APCI system, an HPLC-MS interface for analyzing samples not suited for ElectroSpray ionization. Its higher power design rapidly heats the nebulizer plume, ensuring efficient droplet evaporation for optimal analytical performance.
  • Higher power heater design for atmospheric pressure chemical ionization.
  • Heats nebulizer plume efficiently for rapid droplet evaporation.
  • Supports 100% organic or aqueous mobile phases, 0.2 to 2.0 mL/min.
  • Ideal for standard analytical column LC–MS, normal and reverse phase.
  • APCI probe temperature adjustable up to 650°C.
  • Optimizes temperature for maximum sensitivity.
  • Aids chemical contamination removal from probe tip.