Learn More
Waters Corp APCI Probe Adaptor Assembly, Mechanical, MS Systems
Catalog No. 502392850 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The APCI Probe Adaptor Assembly is a replacement part designed to facilitate the use of Tool Free Probes. This mechanical component is specifically used with MS (Mass Spectrometry) systems.
  • Product type: Spare parts kits
  • Units per package: 1 pk
  • Type: Mechanical
  • System type: MS
  • Replacement schedule: As required
Catalog No. 50-239-2850 Supplier Waters Corp Supplier No. 700011243
The APCI Probe Adaptor Assembly is a replacement part designed to facilitate the use of Tool Free Probes. This mechanical component is specifically used with MS (Mass Spectrometry) systems.
  • Product type: Spare parts kits
  • Units per package: 1 pk
  • Type: Mechanical
  • System type: MS
  • Replacement schedule: As required