Learn More
Waters Corp Waters Corporation APCI Probe Service Kit, MS System, Rebuild Kit
Catalog No. 50179753 Shop All Waters Corporation Products
Encompass_Preferred
The APCI Probe Service Kit contains all the consumable parts required to maintain the APCI probe on a regular basis.
  • Contains all consumable parts required for regular maintenance of the APCI probe.
  • Functions as a rebuild kit.
  • Suitable for MS systems.
  • Replacement recommended as required.
  • Includes one unit per package.
Catalog No. 50-179-753 Supplier Waters Corp Supplier No. 700004673
The APCI Probe Service Kit contains all the consumable parts required to maintain the APCI probe on a regular basis.
  • Contains all consumable parts required for regular maintenance of the APCI probe.
  • Functions as a rebuild kit.
  • Suitable for MS systems.
  • Replacement recommended as required.
  • Includes one unit per package.