Learn More
TARGETMOL CHEMICALS INC APCIN 10MG
SDP
Catalog No. 502579060
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Apcin is a potent and competitive inhibitor of anaphase-promoting complex/cyclosome (APC/C(Cdc20)) E3 ligase activity. Apcin competitively inhibits APC/C-dependent ubiquitylation by binding to Cdc20 and preventing substrate recognition. It acts by blocking mitotic exit and being synergistically amplified by co-addition of Ts-Arg-OMe. purity: 96%
Catalog No. 50-257-9060 Supplier TARGETMOL CHEMICALS INC Supplier No. T856110MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Apcin is a potent and competitive inhibitor of anaphase-promoting complex/cyclosome (APC/C(Cdc20)) E3 ligase activity. Apcin competitively inhibits APC/C-dependent ubiquitylation by binding to Cdc20 and preventing substrate recognition. It acts by blocking mitotic exit and being synergistically amplified by co-addition of Ts-Arg-OMe. purity: 96%